To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:G-Protein Subunit beta 3
- Antigen Symbol:GNB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Epitope:Human GNB3 amino acids 216-230.
- Cross Adsorption:Yes
- Format:Liquid
- Antigen Synonyms:Transducin beta chain 3|Guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-3|GNB3
- Immunogen:Human GNB3 peptide (216-230 aa)
- Cat. No.:10781-938
- Supplier no.:C-1520-100
G-Protein beta3 (GNB3) is a guanine nucleotide-binding protein (G protein). G-proteins are involved as a modulator or transducer in various transmembrane signaling systems (Ref: SWISSPROT).
The antibody reacts specifically with GNB3 protein of human, mouse, and rat. Because of homology it is expects to react with other mammalian species as well. The antibody detects a prominent band ~38 kDa in western blot with extracts from human adipose tissue.
Application Information:
WB. Suggested dilution at 1:1,000 to 1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: G-Protein subunit beta-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human GNB3 amino acids 216-230.
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat