To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:REST Corepressor 1
- Antigen Symbol:RCOR1
- Clonality:Monoclonal
- Clone:S72-8
- Conjugation:ATTO 488
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:23186
- Antigen Synonyms:COREST|RCOR|KIAA0071
- Amino Acid Number:109 to 293
- Storage Buffer:PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
- Storage Temperature:Store at 4°C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant fusion protein corresponding to amino acids of 109-293, including ELM2 and SANT domains, of human RCOR1.
- Purification:Protein G Purification
- Cat. No.:10734-190
- Supplier no.:MAB11928
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant RCOR1.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: RCOR1
Clonality: Monoclonal
Clone: S72-8
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0