To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:SH3-Domain GRB2-like 1
- Antigen Symbol:SH3GL1
- Clonality:Monoclonal
- Clone:S51-1
- Conjugation:AP (Alkaline Phosphatase)
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:81922
- Antigen Synonyms:SH3P8|SH3D2B|EEN|CNSA1
- Amino Acid Number:287 to 302
- Storage Buffer:PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
- Storage Temperature:Store at 4°C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids of 287-302 of rat Sh3gl1.
- Purification:Protein G Purification
- Cat. No.:10738-182
- Supplier no.:MAB11881
Specifications
About this item
Mouse monoclonal antibody raised aginst synthetic peptide of Sh3gl1.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: SH3GL1
Clonality: Monoclonal
Clone: S51-1
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: