To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Succinate Dehydrogenase Complex, Subunit D, Integral Membrane Protein
- Antigen Symbol:SDHD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:6392
- Antigen Synonyms:cybS|PGL|CII-4|CWS3|QPs3|PGL1|CBT1|SDH4
- Storage Buffer:PBS (0.02% sodium azide)
- Storage Temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region of human SDHD.
- Purification:Affinity purification
- Cat. No.:10737-454
- Supplier no.:PAB25486
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SDHD.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunocytochemistry: 2.5 ?g/ml
Type: Primary
Antigen: SDHD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat