To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:V-rel Avian Reticuloendotheliosis Viral Oncogene homolog
- Antigen Symbol:REL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5966
- Antigen Synonyms:4.1-G|c-Rel|4.1G
- Modification:phospho-Ser503
- Storage Buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
- Sequence:T-S-Sp-D-S
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S503 of human REL.
- Purification:purified by Affinity Chromatography
- Cat. No.:10675-262
- Supplier no.:PAB25241
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of REL.
Recommended Dilutions: Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: REL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human