To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Transglutaminase 7
- Antigen Symbol:TGM7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:116179
- Antigen Synonyms:TGMZ|TGM7|Protein-glutamine gamma-glutamyltransferase Z|Transglutaminase Z|TGZ
- Amino Acid Number:1 to 710
- Storage Buffer:1x PBS, pH 7.4
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:TGM7 (AAI53106.1, 1 a.a. ~ 710 a.a) full-length human protein.
- Cat. No.:10574-894
- Supplier no.:H00116179-B01P
Mouse polyclonal antibody raised against a full-length human TGM7 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: TGM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human