Order Entry
United States
ContactUsLinkComponent
 
Anti-C1RL Mouse Polyclonal Antibody
  10552-030
 :  Abnova
undefined
Anti-C1RL Mouse Polyclonal Antibody
  10552-030
 :  Abnova
 :  H00051279-B01P
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Complement Component 1, r subcomponent-like
  • Antigen Symbol:
    C1RL
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    51279
  • Antigen Synonyms:
    CLSPa|C1r-LP|C1RL1|C1RLP
  • Amino Acid Number:
    1 to 487
  • Storage Buffer:
    1x PBS, pH 7.4
  • Sequence:
    MPGPRVWGKYLWRSPHSKGCPGAMWWLLLWGVLQACPTRGSVLLAQELPQQLTSPGYPEPYGKGQESSTDIKAPEGFAVRLVFQDFDLEPSQDCAGDSVTISFVGSDPSQFCGQQGSPLGRPPGQREFVSSGRSLRLTFRTQPSSENKTAHLHKGFLALYQTVAVNYSQPISEASRGSEAINAPGDNPAKVQNHCQEPYYQAAAAGALTCATPGTWKDRQDGEEVLQCMPVCGRPVTPIAQNQTTLGSSRAKLGNFPWQAFTSIHGRGGGALLGDRWILTAAHTVYPKDSVSLRKNQSVNVFLGHTAIDEMLKLGNHPVHRVVVHPDYRQNESHNFSGDIALLELQHSIPLGPNVLPVCLPDNETLYRSGLLGYVSGFGMEMGWLTTELKYSRLPVAPREACNAWLQKRQRPEVFSDNMFCVGDETQRHSVCQGDSGSVYVVWDNHAHHWVATGIVSWGIGCGEGYDFYTKVLSYVDWIKGVMNGKN
  • Storage Temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    C1RL (AAI56781.1, 1 a.a. ~ 487 a.a) full-length human protein.
  • Cat. No.:
    10552-030
  • Supplier no.:
    H00051279-B01P

 

 

Mouse polyclonal antibody raised against a full-length human C1RL protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: C1RL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human