To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Tumor Necrosis Factor Receptor Superfamily, Member 10A
- Antigen Symbol:TNFRSF10A
- Clonality:Monoclonal
- Clone:32A1380
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:8797
- Antigen Synonyms:TRAILR1|DR4|APO2|CD261|TRAILR-1
- Amino Acid Number:1 to 20
- Storage Buffer:PBS (0.05% BSA, 0.05% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 1-20 of human TNFRSF10A.
- Cat. No.:10742-022
- Supplier no.:MAB0054
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of TNFRSF10A.
Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: 32A1380
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human