To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:ALX Homeobox 4
- Antigen Symbol:ALX4
- Clonality:Monoclonal
- Clone:KAbB4
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:60529
- Antigen Synonyms:CRS5|KIAA1788|FND2
- Storage Buffer:PBS (0.08% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Human ALX4.
- Cat. No.:10688-900
- Supplier no.:MAB1834
Mouse monoclonal antibody raised against ALX4.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: ALX4
Clonality: Monoclonal
Clone: KAbB4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human