To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Lymphoid Enhancer-Binding Factor 1
- Antigen Symbol:LEF1
- Clonality:Monoclonal
- Clone:REMB1
- Conjugation:Unconjugated
- Host:
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:51176
- Antigen Synonyms:TCF1ALPHA|TCF1-alpha|DKFZp586H0919|LEF-1|LEF1
- Storage Buffer:PBS (0.08% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length LEF1.
- Cat. No.:10723-482
- Supplier no.:MAB1943
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant LEF1.
Recommended Dilutions: Western Blot: 1 to 5 ?g/ml; Immunofluorescence on paraformaldehyde fixed cells (1:1000 or 1 ?g/ml
Type: Primary
Antigen: LEF1
Clonality: Monoclonal
Clone: REMB1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human, Mouse