To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin beta 1
- Antigen Symbol:ITGB1
- Clonality:Monoclonal
- Clone:k2D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:3688
- Antigen Synonyms:FNRB|integrin VLA-4 beta subunit|VLA-BETA|VLAB|VLA-4 subunit beta|CD29|beta polypeptide|MDF2|MSK12|GPIIA|Fibronectin receptor subunit beta|very late activation protein
- Amino Acid Number:34 to 141
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 34-141 of human ITGB1.
- Cat. No.:10719-790
- Supplier no.:MAB1129
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant ITGB1.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: k2D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human