To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 2 (P49/P100)
- Antigen Symbol:NFKB2
- Clonality:Monoclonal
- Clone:9D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:4791
- Antigen Synonyms:H2TF1|p52|LYT-10|p100|NF-kB2|CVID10|LYT10
- Amino Acid Number:1 to 454
- Storage Buffer:HEPES, 150 mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant His fusion protein corresponding to amino acids 1-454 of human NFKB2.
- Purification:Ammonium sulfate precipitation
- Cat. No.:10726-582
- Supplier no.:MAB1005
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinantNFKB2.
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: NFKB2
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 9D2
Epitope:
Host: IgG2b kappa
Isotype: Mouse
Reactivity: Human, Mouse, Rat