To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Autophagy Related 10
- Antigen Symbol:ATG10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:83734
- Antigen Synonyms:DKFZP586I0418|pp12616|FLJ13954|APG10L|ATG10
- Storage Buffer:PBS
- Storage Temperature:Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with Blue Carrier Protein) corresponding to C-terminus of human ATG10.
- Cat. No.:10690-336
- Supplier no.:PAB0732
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ATG10.
Recommended Dilutions: Immunohistochemistry: 1:100-1:1000; Immunofluorescence: 1:100-1:1000
Type: Primary
Antigen: ATG10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human