To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 9 Member 8
- Antigen Symbol:SLC9A8
- Clonality:Monoclonal
- Clone:7A11
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:23315
- Antigen Synonyms:NHE-8|NHE8
- Storage Buffer:Tris-glycine, 150 mM NaCl (0.05% sodium azide)
- Storage Temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to C-terminus of human SLC9A8.
- Cat. No.:10737-970
- Supplier no.:MAB2292
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant SLC9A8.
Recommended Dilutions: Western Blot: 2 ?g/ml
Type: Primary
Antigen: SLC9A8
Clonality: Monoclonal
Clone: 7A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human