To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Fibroblast Growth Factor 8
- Antigen Symbol:FGF8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:2253
- Antigen Synonyms:KAL6|FGF-8|HH6|AIGF|HBGF-8
- Storage Buffer:0.9 mg NaCl, 0.2 mg Na2HPO4 (5 mg BSA, 0.05 mg sodium azide, 0.05 mg Thimerosal)
- Storage Temperature:Store at –20 °C on dry atmosphere. After reconstitution with 200 ?l of deionized water and concentration will be 500 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at C-terminus of human FGF8.
- Purification:Affinity purification
- Cat. No.:10720-274
- Supplier no.:PAB12306
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of FGF8.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-2 ?g/ml
Type: Primary
Antigen: FGF8
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope: C-terminal
Host:
Isotype: Rabbit
Reactivity: Human, Mouse, Rat