To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cadherin 15
- Antigen Symbol:CDH15
- Clonality:Monoclonal
- Clone:21G4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:1013
- Antigen Synonyms:CDH15|Muscle Cadherin|M-Cadherin|CDH14|Cadherin-14|Cadherin-15|CDH3
- Storage Buffer:2x PBS (PEG, sucrose, 0.09% sodium azide)
- Storage Temperature:Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to extracellular region of human CDH15.
- Cat. No.:10707-240
- Supplier no.:MAB3922
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CDH15.
Recommended Dilutions: ELISA: 0.05 ?g/ml; Western Blot: 1 ?g/ml for AP/BCIP/NBT (MTT) detection 0.5 ?g/ml for HRP/ECL detection; Immunofluorescence: 5 ?g/ml
Type: Primary
Antigen: CDH15
Clonality: Monoclonal
Clone: 21G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: