Order Entry
United States
ContactUsLinkComponent
Anti-HBcAg Monoclonal Antibody [clone: 13A9]
Anti-HBcAg Monoclonal Antibody [clone: 13A9]
Catalog # 10720-850
Supplier:  Abnova
Anti-HBcAg Monoclonal Antibody [clone: 13A9]
Catalog # 10720-850
Supplier:  Abnova
Supplier Number:  MAB4774
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Hepatitis B Core Antigen
  • Antigen Symbol:
    HBcAg
  • Clonality:
    Monoclonal
  • Clone:
    13A9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Virus
  • Size:
    100 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Antigen Synonyms:
    Hepatitis B virus nucleocapsid particles|HepB
  • Storage Buffer:
    PBS, pH 7.4
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to hepatitis B core antigen.
  • Cat. No.:
    10720-850
  • Supplier no.:
    MAB4774

Specifications

About this item

Mouse monoclonal antibody raised against recombinant hepatitis B core antigen.

Type: Primary
Antigen: Hepatitis B virus core antigen
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 13A9
Epitope:
Host: IgG2b
Isotype:
Reactivity: Viruses