To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:NOP58 ribonucleoprotein homolog
- Antigen Symbol:NOP58
- Clonality:Monoclonal
- Clone:37C12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Yeast
- Size:200 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:854487|850894
- Storage Buffer:PBS (50% glycerol, <0.1% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified NOP58/SIK1 from yeast nuclear protein preparations.
- Cat. No.:10676-926
- Supplier no.:MAB3346
Mouse monoclonal antibody raised against native NOP58/SIK1.
Recommended Dilutions: Immunocytochemistry: 1:500-1:5000; Immunofluorescence: 1:500-1:5000
Type: Primary
Antigen: NOP58
Clonality: Monoclonal
Clone: 37C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Yeast