To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Transforming Growth Factor Beta 1
- Antigen Symbol:TGFB1
- Clonality:Monoclonal
- Clone:TB21
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:7040
- Antigen Synonyms:CED|TGFbeta|TGFB|DPD1|LAP
- Storage Buffer:PBS
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human TGFB1.
- Cat. No.:10741-080
- Supplier no.:MAB2505
Mouse monoclonal antibody raised against native TGFB1.
Recommended Dilutions: Western Blot: 5-20 ng/ml; Immunohistochemistry: 1:1000
Type: Primary
Antigen: TGFB1
Clonality: Monoclonal
Clone: TB21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat