To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:NCK Adaptor Protein 2
- Antigen Symbol:NCK2
- Clonality:Monoclonal
- Clone:AF3F11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:8440
- Antigen Synonyms:growth factor receptor-bound protein 4|SH2/SH3 adaptor protein NCK-beta|noncatalytic region of tyrosine kinase beta|GRB4|cytoplasmic protein NCK2|NCKbeta
- Storage Buffer:HEPES, 150 mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant His fusion protein corresponding to human NCK2.
- Purification:Ammonium sulfate precipitation
- Cat. No.:10726-738
- Supplier no.:MAB2727
Mouse monoclonal antibody raised against recombinant NCK2.
Recommended Dilutions: Western Blot: 1:2000; Immunoprecipitation: 1 ?l
Type: Primary
Antigen: NCK2
Clonality: Monoclonal
Clone: AF3F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat