Order Entry
United States
ContactUsLinkComponent
Anti-SPINK4 Rabbit Polyclonal Antibody
Anti-SPINK4 Rabbit Polyclonal Antibody
Catalog # 10739-310
Supplier:  Abnova
undefined
Anti-SPINK4 Rabbit Polyclonal Antibody
Catalog # 10739-310
Supplier:  Abnova
Supplier Number:  PAB20369
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Serine Protease Inhibitor Kazal-Type 4
  • Antigen Symbol:
    SPINK4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    27290
  • Storage Buffer:
    PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
  • Sequence:
    LPFSRMPICEHMVESPTCSQMSNLVCGTDGLTYTNECQLCLARIKTKQDIQIMKDGK
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids of human SPINK4.
  • Purification:
    Antigen affinity purification
  • Cat. No.:
    10739-310
  • Supplier no.:
    PAB20369

Specifications

About this item

Rabbit polyclonal antibody raised against recombinant SPINK4.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: SPINK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human