Order Entry
United States
ContactUsLinkComponent
Anti-NLRP3 Mouse Monoclonal Antibody [clone: Nalpy3-b]
Anti-NLRP3 Mouse Monoclonal Antibody [clone: Nalpy3-b]
Catalog # 10733-044
Supplier:  Abnova
Anti-NLRP3 Mouse Monoclonal Antibody [clone: Nalpy3-b]
Catalog # 10733-044
Supplier:  Abnova
Supplier Number:  MAB3724
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    NLR Family, pyrin Domain Containing 3
  • Antigen Symbol:
    NLRP3
  • Clonality:
    Monoclonal
  • Clone:
    Nalpy3-b
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    114548
  • Antigen Synonyms:
    MWS|AII|LRR and PYD domains-containing protein 3|FCU|AGTAVPRL|cryopyrin|NACHT|and PYD-containing protein 3|NALP3|cold autoinflammatory syndrome 1 protein|C1orf7|CLR1.1|AVP|leucine-rich repeat-|CIAS1|nucleotide-binding oligomerization domain|FCAS|PYRIN-containing APAF1-like protein 1|leucine rich repeat and pyrin domain containing 3|PYPAF1|caterpiller protein 1.1|LRR and PYD containing protein 3|NACHT domain-
  • Storage Buffer:
    PBS (0.02% sodium azide)
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human NLRP3.
  • Cat. No.:
    10733-044
  • Supplier no.:
    MAB3724

Specifications

About this item

Mouse monoclonal antibody raised against recombinant NLRP3.

Recommended Dilutions: Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: NLRP3
Clonality: Monoclonal
Clone: Nalpy3-b
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human