To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Mycobacterium tuberculosis RV2623
- Antigen Symbol:Mycobacterium tuberculosis RV2623
- Clonality:Monoclonal
- Clone:E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Bacteria
- Western Blot:Yes
- Size:200 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at 4°C.
- Immunogen:Recombinant protein corresponding to Mycobacterium tuberculosis RV2623.
- Cat. No.:10677-184
- Supplier no.:MAB3173
Specifications
About this item
Mouse monoclonal antibody raised against recombinant Mycobacterium tuberculosis RV2623.
Type: Primary
Antigen: Mycobacterium tuberculosis RV2623
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: E5
Epitope:
Host: IgG2b
Isotype: Mouse
Reactivity: Bacteria