To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:FK506 Binding Protein 5
- Antigen Symbol:FKBP5
- Clonality:Monoclonal
- Clone:Hi51B
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Hamster,Rabbit,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2289
- Antigen Synonyms:AIG6|FKBP51|P54|FKBP54|PPIase|Ptg-10
- Storage Buffer:PBS (0.09% sodium azide, 50% glycerol)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human FKBP5.
- Cat. No.:10714-814
- Supplier no.:MAB6633
Mouse monoclonal antibody raised against synthetic peptide of FKBP5.
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: FKBP5
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: Hi51B
Epitope:
Host: IgG
Isotype: Mouse
Reactivity: Dog, Hamster, Human, Mouse, Rabbit, Rat