To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Acetyl-CoA Acetyltransferase 2
- Antigen Symbol:ACAT2
- Clonality:Monoclonal
- Clone:AT2.G7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:39
- Storage Buffer:PBS, pH 7.4 (0.05% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to N-terminus of human ACAT2.
- Purification:Protein A purification
- Cat. No.:10687-810
- Supplier no.:MAB8767
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant ACAT2.
Recommended Dilutions: ELISA: 1:2000-5000; Western Blot: 1:1000
Type: Primary
Antigen: ACAT2
Clonality: Monoclonal
Clone: AT2.G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human