To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Thomsen Friedenreich Tn Antigen
- Antigen Symbol:Thomsen Friedenreich Tn Antigen
- Clonality:Monoclonal
- Clone:Tn 218
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM kappa
- Reactivity:Sheep,Human
- Size:500 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS, pH 7.4 (0.05% sodium azide)
- Storage Temperature:Store at 4°C.
- Immunogen:Asialo ovine submaxillary mucin (AOSM).
- Cat. No.:10736-178
- Supplier no.:MAB6198
Specifications
About this item
Mouse monoclonal antibody raised against Thomsen Friedenreich Tn antigen.
Recommended Dilutions: Immunohistochemistry (Frozen sections): 1:50-1:100; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:50-1:100
Type: Primary
Antigen: Thomsen Friedenreich Tn antigen
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: Tn 218
Epitope:
Host: IgM kappa
Isotype: Mouse
Reactivity: Human, Sheep