To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:TIP41 toR signaling pathway Regulator-like
- Antigen Symbol:TIPRL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross Adsorption:No
- Format:Liquid
- Gene ID:261726
- Antigen Synonyms:TOR signalling pathway regulator-like|TIP|dJ69E11.3|TIP41|TIPRL|MGC3794|putative MAPK activating protein|CG9578-like|TOR signaling pathway regulator-like (S. cerevisiae)|yeast YPR037W and worm C02C2.6 predicted proteins-like|OTTHUMP00000032581|OTTHUMP00000032582
- Storage Buffer:Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
- Sequence:C-RIDPNPADSQKSTQ
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at C-terminus of human TIPRL.
- Purification:Antigen affinity purification
- Cat. No.:10741-328
- Supplier no.:PAB18749
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of TIPRL.
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: TIPRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Goat
Isotype:
Reactivity: Human