Anti-TGFA Rabbit Polyclonal Antibody
Catalog # 10741-076
Supplier: Abnova
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Transforming Growth Factor, Alpha
- Antigen Symbol:TGFA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:7039
- Antigen Synonyms:transforming growth factor alpha|TGF-alpha|TGFalpha|ETGF|TGF-a|TGF type 1|TGF alpha|EGF-like TGF
- Storage Buffer:PBS.
- Storage Temperature:Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at 4 °C for one month. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic protein corresponding to human TGFA.
- Purification:Protein G purification
- Cat. No.:10741-076
- Supplier no.:PAB19555
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic protein of TGFA.
- Recommended Dilutions: ELISA: 1-2 μg/ml; Western Blot: 2-10 μg/ml; Immunohistochemistry: 2-10 μg/ml
Type: Primary
Antigen: TGFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat