To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Interleukin 1 Alpha
- Antigen Symbol:IL1A
- Clonality:Monoclonal
- Clone:5G3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:3552
- Antigen Synonyms:Hematopoietin-1|IL-1 alpha|pro-interleukin-1-alpha|IL1A|preinterleukin 1 alpha|hematopoietin-1|interleukin-1 alpha
- Storage Buffer:Citrate-Tris-HCl buffer, pH 7.0 (0.02% Procl300)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human IL1A.
- Purification:Affinity purification
- Cat. No.:10719-102
- Supplier no.:MAB9932
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant IL1A.
Recommended Dilutions: Western Blot: 1:1000; Immunohistochemistry: 1:10
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: 5G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human