To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Shigella dysenteriae IpaA
- Antigen Symbol:Shigella dysenteriae IpaA
- Clonality:Monoclonal
- Clone:4f44s
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Amino Acid Number:240 to 581
- Storage Buffer:citrate-Tris-HCl, pH7.0 (0.02% Procl300)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 240-581 of Shigella dysenteriae IpaA.
- Purification:Affinity purification
- Cat. No.:10732-164
- Supplier no.:MAB8947
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant Shigella dysenteriae IpaA.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: Shigella dysenteriae IpaA
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 4f44s
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: