To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Defensin, beta 103
- Antigen Symbol:DEFB103B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:55894|414325
- Antigen Synonyms:HBD3|HBP3|DEFB-3|BD-3|HBP-3|HBD-3|DEFB103|DEFB3
- Storage Buffer:PBS, pH 7.4 (0.02% sodium azide)
- Storage Temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Full length human DEFB103A/DEFB103B.
- Cat. No.:10710-442
- Supplier no.:PAB17105
Specifications
About this item
Rabbit polyclonal antibody raised against DEFB103A/DEFB103B.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: DEFB103B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human