To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cytochrome P450, Family 26, Subfamily B, Polypeptide 1
- Antigen Symbol:CYP26B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:56603
- Antigen Synonyms:Cytochrome P450 26B1|Retinoic acid-metabolizing cytochrome|RHFCA|P450 26A2|P450RAI2|P450 retinoic acid-inactivating 2|P450RAI-2|CYP26A2
- Storage Buffer:Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
- Sequence:C-SQARSEDKDGRFH
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human CYP26B1.
- Purification:Antigen affinity purification
- Cat. No.:10709-826
- Supplier no.:PAB7139
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of CYP26B1.
Recommended Dilutions: ELISA: 1:128000
Type: Primary
Antigen: CYP26B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: