Order Entry
United States
ContactUsLinkComponent
Anti-CXCR6 Rabbit Polyclonal Antibody
Anti-CXCR6 Rabbit Polyclonal Antibody
Catalog # 10683-744
Supplier:  Abnova
undefined
Anti-CXCR6 Rabbit Polyclonal Antibody
Catalog # 10683-744
Supplier:  Abnova
Supplier Number:  PAB26078
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Chemokine (C-X-C motif) Receptor 6
  • Antigen Symbol:
    CXCR6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Gorilla,
    Chimpanzee,
    Human
  • Size:
    50 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    10663
  • Antigen Synonyms:
    TYMSTR|C-X-C chemokine receptor type 6|CD186|BONZO|STRL33|Bonzo|CDw186|CXC-R6
  • Storage Buffer:
    PBS (0.1% sodium azide)
  • Storage Temperature:
    Store at 4°C. For long term storage store at –80 °C . Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 16 amino acids at C-terminus of human CXCR6.
  • Purification:
    Immunoaffinity chromatography
  • Cat. No.:
    10683-744
  • Supplier no.:
    PAB26078

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of CXCR6.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 19 ?g/ml

Type: Primary
Antigen: CXCR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: