Order Entry
United States
ContactUsLinkComponent
 
Anti-NSD1 Mouse Monoclonal Antibody [clone: 1C12]
  10595-432
 :  Abnova
undefined
Anti-NSD1 Mouse Monoclonal Antibody [clone: 1C12]
  10595-432
 :  Abnova
 :  H00064324-M05
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Nuclear Receptor binding SET Domain Protein 1
  • Antigen Symbol:
    NSD1
  • Clonality:
    Monoclonal
  • Clone:
    1C12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Gene ID:
    64324
  • Antigen Synonyms:
    Lysine N-methyltransferase 3B|ARA267|H3-K36-HMTase|H3 lysine-36 and H4 lysine-20 specific|Histone-lysine N-methyltransferase|NSD1|KMT3B|NR-binding SET domain-containing protein|Nuclear receptor-binding SET domain-containing protein 1|H4-K20-HMTase|Androgen receptor-associated protein of 267 kDa|Androgen receptor coactivator 267 kDa protein
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    10595-432
  • Supplier no.:
    H00064324-M05

 

 

Type: Primary
Antigen: NSD1
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human