Anti-IL4R Mouse Polyclonal Antibody
10561-708
: Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Interleukin 4 Receptor
- Antigen Symbol:IL4R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Gene ID:3566
- Antigen Synonyms:IL-4 receptor subunit alpha|IL4-BP|IL4R|IL-4-binding protein|ORF Names: 582J2.1|Soluble IL-4 receptor subunit alpha|IL-4RA|IL-4R-alpha|Interleukin-4 receptor subunit alpha|IL-4R subunit alpha|IL4RA|Soluble IL-4R-alpha|sIL4Ralpha/prot|Soluble interleukin-4 receptor subunit alpha|CD124
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10561-708
- Supplier no.:H00003566-B01
Type: Primary
Antigen: IL4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human