Order Entry
United States
ContactUsLinkComponent
Anti-SLC25A5 Rabbit Polyclonal Antibody
Anti-SLC25A5 Rabbit Polyclonal Antibody
Catalog # 10601-592
Supplier:  Abnova
undefined
Anti-SLC25A5 Rabbit Polyclonal Antibody
Catalog # 10601-592
Supplier:  Abnova
Supplier Number:  H00000292-D03
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Solute Carrier Family 25 Member A5
  • Antigen Symbol:
    SLC25A5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Gene ID:
    292
  • Antigen Synonyms:
    N-terminally processed|ADP/ATP translocase 2|ANT 2|Adenine nucleotide translocator 2|DP/ATP translocase 2|ADP,ATP carrier protein 2|fibroblast isoform|Solute carrier family 25 member 5|ANT2|ADP,ATP carrier protein
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    10601-592
  • Supplier no.:
    H00000292-D03

Specifications

About this item

Type: Primary
Antigen: SLC25A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human