ContactUsLinkComponent
Quantitative Analysis
Quantitative Analysis
Catalog # 470163-084
Supplier:  Ward's Science
undefined
Quantitative Analysis
Catalog # 470163-084
Supplier:  Ward's Science
Supplier Number:  IS8009
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Quantitative Analysis
  • Environmentally Preferable:
  • No. of lab groups:
    15
  • Cat. No.:
    470163-084
  • Includes:
    Teacher's Guide and Student Copymaster
  • Supplier no.:
    IS8009
Specifications
About this item
Analytical Chemistry for AP Students
  • Classic AP Chemistry Experiment
  • Instructions Included
Colorimetric or turbidimetric methods of analysis have been developed for most of the elements and ionic species. These methods can be employed to quickly determine the chemical composition of an unknown substance. Before attempting to analyze an unknown it is useful to develop a logical process to follow. Care must be taken in sampling, collecting and washing precipitates, performing confirmatory tests and most importantly, recording data. It is also good laboratory practice to always analyze a known with each unknown as a double check on the reagents being used. The unknowns will be analyzed for one of six possible anions: Cl-, Br-, I-, PO42-, NO3-, and SO42-. A standard solution for each anion will also be run.
Includes:
Sodium Bromide Solution
Sodium Chloride Solution
Sodium Iodide Solution
Sodium Nitrate Solution
Sodium Phosphate Solution
Sodium Sulfate Solution
Ammonium Hydroxide
Mineral Oil
Ammonium Molybdate Solution
Diphenylamine
Barium Chloride Solution
Potassium Permanganate Solution
Ferric Nitrate Solution
Silver Nitrate Solution