Anti-PI 4 Kinase type 2 beta Rabbit Polyclonal Antibody
Catalog # 80084-720
Supplier: Abgent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:PI 4 Kinase type 2 beta
- Antigen Symbol:PI4K2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Environmentally Preferable:
- Epitope:C-Terminal (319–336 aa)
- Cross Adsorption:No
- Antigen Synonyms:Type II phosphatidylinositol 4-kinase beta isoform|PI4K2B protein|Phosphatidylinositol 4-kinase type-II beta
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80084-720
- Supplier no.:AP8033B
Specifications
About this item
Type: Primary
Antigen: PI 4 Kinase type 2 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse