To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:RNA Binding Protein, Fox-1 homolog 1
- Antigen Symbol:A2BP1
- Clonality:Monoclonal
- Clone:D8H8
- Conjugation:DyLight® 488
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:54715
- Antigen Synonyms:FOX1 hexaribonucleotide binding protein 1|2BP1|RNA binding protein fox-1 homolog 1|HRNBP1 ataxin 2-binding protein 1|RNA binding protein|Fox-1 homolog A|A2BP1 Hexaribonucleotide-binding protein 1|A2BP|fox-1 homolog (C. elegans) 1|Ataxin-2-binding protein 1|FOX-1
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:Mouse recombinant A2BP1 [Swiss-Prot# Q9JJ43]
- Purification:Protein G purified
- Cat. No.:102715-782
- Supplier no.:NBP2-13169G
Specifications
About this item
The A2BP1 Antibody (D8H8) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to A2BP1. This antibody reacts with human, mouse. The A2BP1 Antibody (D8H8) [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: A2BP1
Clonality: Monoclonal
Clone: D8H8
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse