To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:NLR family, CARD domain containing 4
- Antigen Symbol:CARD12
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 μl
- Environmentally Preferable:
- Gene ID:58484
- Antigen Synonyms:Ipaf|CLAN1ipaf|caspase recruitment domain family|and NACHT-containing protein|CARD domain containing 4|CLR2.1|CARD12leucine rich repeat and CARD domaincontaining 4|member 12|NLR family|Caspase recruitment domain-containing protein 12|Ice protease-activating factor|CLANCARD|LRR|Clan protein
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:A synthetic peptide made to an internal portion of the human CARD12 protein (between residues 600-700) [UniProt Q9NPP4]
- Purification:Immunogen affinity purified
- Cat. No.:102715-550
- Supplier no.:NBP1-78979G
Specifications
About this item
The CARD12 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to CARD12. This antibody reacts with human, mouse. The CARD12 Antibody [DyLight 488] has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CARD12
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse