To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Blooms Syndrome Protein Blm
- Antigen Symbol:BLM
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 ml
- Environmentally Preferable:
- Gene ID:641
- Antigen Synonyms:MGC126616|Bloom syndrome|RecQ protein-like 3|EC 3.6.4.12|RecQ helicase-like|BSBloom syndrome protein|RecQ2|MGC131618|RecQ-like type 2|EC 3.6.1|RECQL3RECQL2|DNA helicase|RECQ2MGC131620
- Modification:p Thr99
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:Synthetic phospho-peptide surrounding the Blooms Syndrome Protein Blm Threonine 99 region [UniProt# P54132]
- Purification:Immunogen affinity purified
- Cat. No.:102715-012
- Supplier no.:NBP1-46851B
The Blooms Syndrome Protein Blm [p Thr99] Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to Blooms Syndrome Protein Blm. This antibody reacts with human. The Blooms Syndrome Protein Blm [p Thr99] Antibody [Biotin] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Blooms Syndrome Protein Blm
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human