About this item
Kit for gel extraction/cleanup of up to 10 g DNA (70 bp to 10 kb) from enzymatic reactions.
- Fast and convenient procedure
- Cleanup of DNA up to 10 kb in three easy steps
- Gel loading dye for convenient sample analysis
- Up to 95% recovery of ready-to-use DNA
This kit provides spin columns, buffers and collection tubes for silica-membrane-based purification of DNA fragments from gels (up to 400 mg slices) or enzymatic reactions. DNA is purified using a simple and fast bind-wash-elute procedure and an elution volume of 30–50 μl. An integrated pH indicator allows easy determination of the optimal pH for DNA binding to the spin column.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: