About this item
Clean and concentrate up to 5 μg DNA with ≥6 μL elution in as little as 2 minutes with 0 μL wash residue carryover.
- 2 minute clean-up protocol with small elution volumes
- Column design allows DNA to be eluted at high concentrations into minimal volumes of water or TE buffer
- Eluted DNA is well suited for use in PCR, DNA sequencing, DNA ligation, endonuclease digestion, RNA transcription, radiolabeling, arrays, etc
About DNA Clean & Concentrator™-5
The DNA Clean & Concentrator™-5 product provides purification of up to 5 µg DNA from PCR, endonuclease digestions, DNA modification reactions, isotope/fluorescence labeling reactions, etc. This product facilitate the removal of DNA polymerases, modifying enzymes, RNA polymerases, ligases, kinases, nucleases, phosphatases, and restriction endonucleases, as well as free dNTPs and their analogs including radiolabeled and fluorescent derivatives. Eluted DNA is suitable for PCR, arrays, ligation, sequencing, etc.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable:
Specifications
Related Information
d4003d
Datasheet
VWR Focus Gene Editing Zymo-Research DNA Clean-Up & Gel DNA Recovery