Order Entry
United States
ContactUsLinkComponent
Anti-Caspase-3 (P17) Rabbit Polyclonal Antibody
Anti-Caspase-3 (P17) Rabbit Polyclonal Antibody
Catalog # 10209-182
undefined
Anti-Caspase-3 (P17) Rabbit Polyclonal Antibody
Catalog # 10209-182
Supplier Number:  PA1961-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Caspase 3
  • Antigen Symbol:
    CASP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Epitope:
    N-Terminal
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Antigen Synonyms:
    procaspase3|Yama|CASP-3|CASP3|CPP32|Apopain|CPP32B|SCA-1|CPP-32|apopain
  • Amino Acid Number:
    67 to 81
  • Sequence:
    TDVDAANLRETFMGL
  • Storage Temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of mouse Caspase-3 (P17) (67-81aa TDVDAANLRETFMGL), different from the related rat sequence by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Cat. No.:
    10209-182
  • Supplier no.:
    PA1961-1

Specifications

About this item

Rabbit IgG polyclonal antibody for Caspase-3(CASP3) detection. Tested with WB, IHC-P in Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Caspase-3 (P17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat