Order Entry
United States
ContactUsLinkComponent
Anti-SHH Rabbit Polyclonal Antibody
Anti-SHH Rabbit Polyclonal Antibody
Catalog # 10207-974
undefined
Anti-SHH Rabbit Polyclonal Antibody
Catalog # 10207-974
Supplier Number:  PA1072-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Sonic Hedgehog
  • Antigen Symbol:
    SHH
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Epitope:
    N-Terminal
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Antigen Synonyms:
    TPT|HLP3|HHG1|SMMCI|HPE3|MCOPCB5|TPTPS
  • Amino Acid Number:
    203 to 218
  • Sequence:
    ATVHLEQGGTKLVKDL
  • Storage Temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human Sonic Hedgehog (203-218aa ATVHLEQGGTKLVKDL), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Cat. No.:
    10207-974
  • Supplier no.:
    PA1072-1

Specifications

About this item

Rabbit IgG polyclonal antibody for Sonic hedgehog protein(SHH) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: SHH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat