To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ras-related protein Rab-5
- Antigen Symbol:RAB5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:RAB5A|Rab5a|RAB 5|RAS|RAB 5A|RAB5|RAS associated protein
- Amino Acid Number:138 to 155
- Sequence:ANKRAVDFQEAQSYADDN
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Rab5 (138-155aa ANKRAVDFQEAQSYADDN), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. No.:10207-270
- Supplier no.:PA1773
Specifications
About this item
Rabbit IgG polyclonal antibody for Ras-related protein Rab-5A(RAB5A) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: RAB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat