Order Entry
United States
ContactUsLinkComponent
Anti-CAR1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-CAR1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # RL20013540100
undefined
Anti-CAR1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # RL20013540100
Supplier Number:  200-1354-0100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CAR1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Size:
    100 µg
  • Environmentally Preferable:
  • Storage Buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Gentamicin Sulfate. Do NOT add Sodium Azide!
  • Concentration:
    1 mg/ml by UV absorbance at 280 nm
  • Cat. No.:
    RL20013540100
  • Supplier no.:
    200-1354-0100

Specifications

About this item

Type: Primary
Antigen: CAR1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human