Bacto™ Peptone, Life Technologies
Catalog # 90000-264
Supplier: Avantor
CAS Number:
Specifications
- Description:Peptone, BD Bacto*
- Size:500 g
- Environmentally Preferable:
- Cat. No.:90000-264
- Supplier no.:211677
Specifications
About this item
Bacto™ Peptone is an animal-origin, enzymatic digest of bovine and porcine animal proteins.
- Animal origin
- Animal containing
- Strong nitrogen source
- Used with a variety of cell types to produce vaccines and biotheraputics
Bacto™ Peptone is an animal-origin, enzymatic digest of bovine and porcine animal proteins. The nutritive value of Bacto™ Peptone is largely dependent on a combination of a negligible quantity of proteases, high concentration of complex constituents and a unique amino acid content that makes this peptone a strong nitrogen source. Customers have successfully used this supplement with a variety of cell types including Saccharomyces cerevisae, Escherichia coli, Pasteurella multocida, Vibro cholera and porcine kidney cells to produce biotherapeutics and vaccines for animal health applications.