Order Entry
United States
ContactUsLinkComponent
 
TGF-beta 1 ELISA Kit, Enzo Life Sciences
Catalog # 89141-224
Supplier:  Avantor
CAS Number:  
undefined
TGF-beta 1 ELISA Kit, Enzo Life Sciences
Catalog # 89141-224
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
  • Environmentally Preferable:
  • Sensitivity:
    3.3pg/mL (range 31.25 - 1000pg/mL)
  • Cat. No.:
    89141-224
  • Supplier no.:
    900-155
  • No. of tests:
    96 wells

Specifications

About this item

For the quantitative determination of TGF-β1 in culture supernatants, plasma, and serum of human, mouse, rat, and cow origin.

The TGF-β1 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450nm. This kit provides ultra-sensitive measurement of as little as 3.3pg/ml TGF-β1 and has been fully validated in activated serum, plasma, and culture supernatants. The ready-to-use liquid color-coded reagents reduce error.

Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Assay buffer 30, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2.