Order Entry
United States
ContactUsLinkComponent
Testosterone ELISA Kit, Enzo Life Sciences
Testosterone ELISA Kit, Enzo Life Sciences
Catalog # 89141-308
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Testosterone ELISA Kit, Enzo Life Sciences
Catalog # 89141-308
Supplier:  Enzo Life Sciences
Supplier Number:  ADI-901-065
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Competitive
  • Host:
  • Primary antibody reactivity:
  • Target protein:
    Testosterone
  • Environmentally Preferable:
  • Time to Results:
    3 hrs
  • Sensitivity:
    5.67 pg/mL (range 7.81 - 2,000 pg/mL)
  • Cat. No.:
    89141-308
  • Supplier no.:
    901-065
  • No. of tests:
    96 wells

Specifications

About this item

For the quantitative determination of testosterone in culture supernatants, plasma, serum, and saliva from any species

The Testosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error. The reproducible results lot-after-lot provide excellent consistency, ideal for longitudinal studies.

Not suitable for human serum and plasma. For those samples use product 89141-348. Cited sample type includes tissue and feces.

Ordering information: Contains GxM Microtiter plate, Conjugate, Antibody, Assay buffer 3, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-112 (1x96 wells).